Gene Transcripts

Gene Symbol
Snx7
Entrez Gene ID
76561
Gene Name
sorting nexin 7
Gene Aliases
2510028H01Rik
Chromosome Location
Chr. 3: 117781497 - 117868936 on Build GRCm38
UniGene ID
Mm.33824
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001190156.1 NP_001177085.1 4 603
NM_029655.3 NP_083931.2 4 603
GenBank mRNA AK011015.1 - 4 578
AK038971.1 - 4 604
AK052487.1 - 4 560
AK170966.1 - 4 550
BC087942.1 - 4 461 30303462

View Full Product Details