Gene Transcripts

Gene Symbol
Cers2
Entrez Gene ID
76893
Gene Name
ceramide synthase 2
Gene Aliases
0610013I17Rik, AI225939, Lass2, TRH3
Chromosome Location
Chr. 3: 95314805 - 95323588 on Build GRCm38
UniGene ID
Mm.181009
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029789.1 NP_084065.1 6, 7 632
GenBank mRNA AK002626.1 - 6, 7 599
AK050161.1 - 6, 7 613
AK146106.1 - 6, 7 611
AK159325.1 - 6, 7 611
AK206786.1 - 2, 3 109
AK216148.1 - 2, 3 86
AY029532.1 - 6, 7 632
BC006847.1 - 6, 7 604 3598473

View Full Product Details