Gene Transcripts

Gene Symbol
Ticrr
Entrez Gene ID
77011
Gene Name
TOPBP1-interacting checkpoint and replication regulator
Gene Aliases
5730590G19Rik, BC028450, Lrrk1
Chromosome Location
Chr. 7: 79660196 - 79698148 on Build GRCm38
UniGene ID
Mm.28132
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029835.1 NP_084111.1 6 1728
GenBank mRNA AK041726.1 - 6 1678
AK051639.1 - 6 1691
AK054492.1 - 6 1678
AK137597.1 - 6 1694
AK157802.1 - 5 3231
BC158103.1 - 6 1692 9088180

View Full Product Details