Gene Transcripts

Gene Symbol
Atg16l1
Entrez Gene ID
77040
Gene Name
autophagy related 16-like 1 (S. cerevisiae)
Gene Aliases
1500009K01Rik, Apg16l, Atg16l, WDR30
Chromosome Location
Chr. 1: 87756006 - 87792428 on Build GRCm38
UniGene ID
Mm.272972
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001205391.1 NP_001192320.1 10 1130
NM_001205392.1 NP_001192321.1 8 1025
NM_029846.4 NP_084122.2 9 1082
GenBank mRNA AB087879.1 - 8 836
AB087880.1 - 9 893
AB087881.1 - 10 941
AB476646.1 - 10 941
AK005181.1 - 10 1055
AK030983.1 - 9 1023
AK131120.1 - 8 1868
AK155358.1 - 8 961
AK163246.1 - 8 964
AK170073.1 - 8 959
AK218161.1 - 4 288
BC049122.1 - 10 1055 6813377

View Full Product Details