Gene Transcripts

Gene Symbol
Bcl9
Entrez Gene ID
77578
Gene Name
B cell CLL/lymphoma 9
Gene Aliases
2610202E01Rik, 8030475K17Rik, A330041G23Rik, Gm130
Chromosome Location
Chr. 3: 97203658 - 97298083 on Build GRCm38
UniGene ID
Mm.226175
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029933.4 NP_084209.3 8 2321
GenBank mRNA AK084676.1 - 1 248
AK147659.1 - 8 2321
AY296061.1 - 5 1837
BC137850.1 - 5 1809 8860869

View Full Product Details