Gene Transcripts

Gene Symbol
Fam53b
Entrez Gene ID
77938
Gene Name
family with sequence similarity 53, member B
Gene Aliases
A930008G19Rik, mKIAA0140
Chromosome Location
Chr. 7: 132712078 - 132813196 on Build GRCm38
UniGene ID
Mm.225649
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175268.4 NP_780477.2 4 517
NM_212473.1 NP_997638.1 4 580
GenBank mRNA AK020830.1 - 4 518
AK031576.1 - 3 361
AK051299.1 - 4 581
AK147350.1 - 4 913
AK147725.1 - 4 709
AK156629.1 - 4 480
AK195672.1 - 3 284
BC035523.1 - 4 512 5400742

View Full Product Details