Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_030098.2 | NP_084374.2 | 2 | 282 | |
GenBank mRNA | AK020595.1 | - | 2 | 226 | |
AK038708.1 | - | 3 | 598 | ||
AY545655.1 | - | 2 | 260 | ||
BC094892.1 | - | 2 | 236 | 6432787 | |
BC132632.1 | - | 2 | 215 | 40130909 | |
BC132634.1 | - | 2 | 214 | 40130911 | |
BY228312.1 | - | 2 | 285 |