Gene Transcripts

Gene Symbol
Plekhj1
Entrez Gene ID
78670
Gene Name
pleckstrin homology domain containing, family J member 1
Gene Aliases
9530063M10Rik
Chromosome Location
Chr. 10: 80796099 - 80798626 on Build GRCm38
UniGene ID
Mm.41479
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_023900.2 NP_076389.2 3, 4 684
GenBank mRNA AK020621.1 - 3, 4 353
AK051734.1 - 3, 4 341
AK150144.1 - 3, 4 299
AK178993.1 - 1, 2 89
AK219000.1 - 1, 2 21
BC005565.1 - 3, 4 282 3966305

View Full Product Details