Gene Transcripts

Gene Symbol
Nipa2
Entrez Gene ID
93790
Gene Name
non imprinted in Prader-Willi/Angelman syndrome 2 homolog (human)
Gene Aliases
2600017P10Rik, 3830408P04Rik, AB041581
Chromosome Location
Chr. 7: 55931266 - 55962655 on Build GRCm38
UniGene ID
Mm.333893
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001256130.1 NP_001243059.1 6 918
NM_001256131.1 NP_001243060.1 5 797
NM_001256132.1 NP_001243061.1 6 722
NM_001256133.1 NP_001243062.1 4 372
NM_023647.6 NP_076136.2 6 1672
GenBank mRNA AB041581.1 - 6 855
AK008106.1 - 6 905
AK011248.1 - 5 742
AK012859.1 - 6 856
AK028734.1 - 6 1630
AK041427.1 - 6 919
AK076051.1 - 6 875
AK076184.1 - 6 859
AK154755.1 - 6 876
BC038499.1 - 6 860 4482150

View Full Product Details