Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001252458.1 | NP_001239387.1 | 17 | 2497 | |
NM_178900.4 | NP_849231.1 | 16 | 2782 | ||
GenBank mRNA | AK037030.1 | - | 16 | 2783 | |
AK154029.1 | - | 16 | 2631 | ||
BC095949.1 | - | 17 | 2447 | 4950957 | |
BC096444.1 | - | 17 | 2465 | 4216827 |