Gene Transcripts

Gene Symbol
Adi1
Entrez Gene ID
104923
Gene Name
acireductone dioxygenase 1
Gene Aliases
AL024210, Fe-ARD, MTCBP-1, Ni-ARD
Chromosome Location
Chr. 12: 28675207 - 28682175 on Build GRCm38
UniGene ID
Mm.291504
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_134052.2 NP_598813.1 3 455
GenBank mRNA AK089966.1 - 3 439
AK145831.1 - 3 432
BC005695.1 - 3 455 3597652

View Full Product Details