Gene Transcripts

Gene Symbol
Brd9
Entrez Gene ID
105246
Gene Name
bromodomain containing 9
Gene Aliases
AL022779
Chromosome Location
Chr. 13: 73937825 - 73960895 on Build GRCm38
UniGene ID
Mm.278509
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK142152.1 - 7 873
AK154645.1 - 4 3113
AK163238.1 - 5 2461
BC031484.1 - 7 871 2654265
BC047372.1 - 7 854
BC057597.1 - 5 2439
BC089379.1 - 7 829

View Full Product Details