Gene Transcripts

Gene Symbol
Pkdcc
Entrez Gene ID
106522
Gene Name
protein kinase domain containing, cytoplasmic
Gene Aliases
AI115348, AW548124, Adtk1, ESTM17, MAd1, Sgk493, Vlk, X83346
Chromosome Location
Chr. 17: 83215283 - 83225069 on Build GRCm38
UniGene ID
Mm.311974
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_134117.2 NP_598878.2 3 1189
GenBank mRNA AB381893.1 - 3 1181
AB436780.1 - 3 1170
BC022157.1 - 2 336
BC086639.1 - 4 1027 6828629

View Full Product Details