Gene Transcripts

Gene Symbol
Tm6sf1
Entrez Gene ID
107769
Gene Name
transmembrane 6 superfamily member 1
Gene Aliases
AI428514, BC023123, C630016D09Rik
Chromosome Location
Chr. 7: 81859001 - 81884079 on Build GRCm38
UniGene ID
Mm.221412
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001291282.1 NP_001278211.1 3 166
NM_145375.4 NP_663350.2 3 674
GenBank mRNA AK046714.1 - 3 575
AK083132.1 - 3 677
AK135100.1 - 3 656
AK136581.1 - 3 167
AK162572.1 - 3 526
AK163621.1 - 3 599
AK172109.1 - 3 590
AK172145.1 - 3 575
BC023123.1 - 3 556 3995280

View Full Product Details