Gene Transcripts

Gene Symbol
Itga1
Entrez Gene ID
109700
Gene Name
integrin alpha 1
Gene Aliases
CD49A, E130012M19Rik, Vla1
Chromosome Location
Chr. 13: 114958079 - 115101964 on Build GRCm38
UniGene ID
Mm.317280
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033228.3 NP_001028400.2 13, 14 2037
GenBank mRNA AK035870.1 - 13, 14 2038
AK040830.1 - 4, 5 461
AK043769.1 - 13, 14 1976
AK137169.1 - 13, 14 1802
BC014765.1 - 13, 14 1901

View Full Product Details