Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_006191.2 | NP_006182.2 | 7 | 1003 | |
GenBank mRNA | AK291796.1 | - | 7 | 756 | |
BC001951.1 | - | 7 | 703 | 2823726 | |
BC007561.1 | - | 7 | 717 | 2988781 | |
BC019222.1 | - | 8 | 676 | ||
BC032111.2 | - | 7 | 684 | ||
BC069786.1 | - | 7 | 656 | 7262559 | |
BC072007.1 | - | 7 | 683 | ||
U59435.1 | - | 7 | 681 | ||
U87954.1 | - | 7 | 779 |