Gene Transcripts

Gene Symbol
Atxn3
Entrez Gene ID
110616
Gene Name
ataxin 3
Gene Aliases
2210008M02Rik, AI463012, AI647473, ATX3, MJD1, Mjd, Sca3, ataxin-3
Chromosome Location
Chr. 12: 101918901 - 101958800 on Build GRCm38
UniGene ID
Mm.271914
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001167914.1 NP_001161386.1 7 652
NM_029705.3 NP_083981.2 7 652
GenBank mRNA AF537188.1 - 7 566
AK008675.1 - 7 611
AK030842.1 - 7 592
BC049743.1 - 7 652
BC087880.1 - 7 647 6771498

View Full Product Details