Gene Transcripts

Gene Symbol
Ppargc1a
Entrez Gene ID
19017
Gene Name
peroxisome proliferative activated receptor, gamma, coactivator 1 alpha
Gene Aliases
A830037N07Rik, ENSMUSG00000079510, Gm11133, PGC-1, PGC-1v, Pgc-1alpha, Pgc1, Pgco1, Ppargc1
Chromosome Location
Chr. 5: 51454249 - 52115958 on Build GRCm38
UniGene ID
Mm.259072
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008904.2 NP_032930.1 13 4771
GenBank mRNA AK032149.1 - 1 1241
AK043827.1 - 1 1275

View Full Product Details