Gene Transcripts

Gene Symbol
KLKB1
Entrez Gene ID
3818
Gene Name
kallikrein B1
Gene Aliases
KLK3, PKK, PKKD, PPK
Chromosome Location
Chr. 4: 186215714 - 186258477 on Build GRCh38
UniGene ID
Hs.237642
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK292624.1 - 5 546
AK313378.1 - 5 495
BC117349.1 - 5 457 40125900
BC117351.1 - 5 457 40125902
M13143.2 - 5 498

View Full Product Details