Gene Transcripts

Gene Symbol
EDN1
Entrez Gene ID
1906
Gene Name
endothelin 1
Gene Aliases
ARCND3, ET1, HDLCQ7, PPET1, QME
Chromosome Location
Chr. 6: 12256464 - 12297194 on Build GRCh38
UniGene ID
Hs.511899
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK226096.1 - 3 560
AK291838.1 - 3 582
AK313914.1 - 3 577
BC009720.1 - 3 514 3900704
BX439515.2 - 3 605
CR541903.1 - 3 311
S56805.1 - 3 647
Y00749.1 - 3 563

View Full Product Details