Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001145457.2 | NP_001138929.2 | 4 | 498 | |
NM_001145458.2 | NP_001138930.2 | 4 | 498 | ||
NM_001242356.2 | NP_001229285.1 | 3 | 213 | ||
NM_001242357.2 | NP_001229286.1 | 3 | 213 | ||
NM_004829.6 | NP_004820.2 | 4 | 498 | ||
XM_011527528.2 | XP_011525830.1 | 4 | 606 | ||
XM_011527529.2 | XP_011525831.1 | 4 | 602 | ||
XM_011527530.2 | XP_011525832.1 | 4 | 594 | ||
GenBank mRNA | AJ001383.1 | - | 4 | 499 | |
AJ006121.1 | - | 4 | 467 | ||
AJ006122.1 | - | 3 | 182 | ||
AJ006123.1 | - | 3 | 182 | ||
AY346373.1 | - | 2 | 139 | ||
BC064806.1 | - | 4 | 485 | 5218848 |