Gene Transcripts
- Gene Symbol
- RABGAP1L
- Entrez Gene ID
- 9910
- Gene Name
- RAB GTPase activating protein 1 like
- Gene Aliases
- HHL, TBC1D18
- Chromosome Location
- Chr. 1: 174159414 - 174995308 on Build GRCh38
- UniGene ID
- Hs.585378
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_014857.4 |
NP_055672.3
|
4, 5 |
817 |
|
XM_005245680.1 |
XP_005245737.1
|
4, 5 |
747 |
|
XM_005245681.2 |
XP_005245738.1
|
4, 5 |
706 |
|
XM_006711693.1 |
XP_006711756.1
|
4, 5 |
747 |
|
XM_011510223.2 |
XP_011508525.1
|
5, 6 |
774 |
|
XM_017002994.1 |
XP_016858483.1
|
4, 5 |
704 |
|
GenBank mRNA |
AB449904.1 |
- |
3, 4 |
540 |
|
AK095838.2 |
- |
4, 5 |
714 |
|
AL157958.1 |
- |
4, 5 |
641 |
|
BC041888.1 |
- |
4, 5 |
710 |
5298493 |
View Full Product Details