Gene Transcripts
- Gene Symbol
- WDSUB1
- Entrez Gene ID
- 151525
- Gene Name
- WD repeat, sterile alpha motif and U-box domain containing 1
- Gene Aliases
- UBOX6, WDSAM1
- Chromosome Location
- Chr. 2: 159235793 - 159286799 on Build GRCh38
- UniGene ID
- Hs.20848
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001128212.2 |
NP_001121684.1
|
7, 8 |
1094 |
|
NM_001128213.1 |
NP_001121685.1
|
7, 8 |
1110 |
|
NM_152528.2 |
NP_689741.2
|
7, 8 |
1002 |
|
XM_005246355.2 |
XP_005246412.2
|
7, 8 |
1088 |
|
XM_006712341.3 |
XP_006712404.1
|
6, 7 |
582 |
|
XM_011510753.2 |
XP_011509055.1
|
7, 8 |
1054 |
|
XM_011510754.2 |
XP_011509056.1
|
7, 8 |
788 |
|
XM_017003481.1 |
XP_016858970.1
|
7, 8 |
1088 |
|
XM_017003483.1 |
XP_016858972.1
|
6, 7 |
909 |
|
GenBank mRNA |
AK093494.1 |
- |
7, 8 |
1002 |
|
AK129983.1 |
- |
7, 8 |
1110 |
|
AL040518.1 |
- |
6, 7 |
478 |
|
View Full Product Details