Gene Transcripts
- Gene Symbol
- POLR3H
- Entrez Gene ID
- 171568
- Gene Name
- RNA polymerase III subunit H
- Gene Aliases
- RPC22.9, RPC8
- Chromosome Location
- Chr. 22: 41525799 - 41544606 on Build GRCh38
- UniGene ID
- Hs.712617
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001018050.3 |
NP_001018060.1
|
3, 4 |
690 |
|
NM_001282884.1 |
NP_001269813.1
|
4, 5 |
418 |
|
NM_001282885.1 |
NP_001269814.1
|
4, 5 |
377 |
|
NM_138338.4 |
NP_612211.1
|
4, 5 |
384 |
|
GenBank mRNA |
AB051452.1 |
- |
3, 4 |
545 |
|
AK026524.1 |
- |
4, 5 |
364 |
|
AY092087.1 |
- |
4, 5 |
362 |
|
BC088367.1 |
- |
3, 4 |
553 |
30385483 |
BU173504.1 |
- |
3, 4 |
332 |
|
CR456459.1 |
- |
4, 5 |
418 |
|
DA449644.1 |
- |
4, 5 |
399 |
|
DA504810.1 |
- |
4, 5 |
366 |
|
View Full Product Details