Gene Transcripts

Gene Symbol
POLR2M
Entrez Gene ID
81488
Gene Name
RNA polymerase II subunit M
Gene Aliases
GCOM1, GRINL1A, Gdown, Gdown1
Chromosome Location
Chr. 15: 57706521 - 57717557 on Build GRCh38
UniGene ID
Hs.437256
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015532.4 NP_056347.1 2 558
GenBank mRNA AF326773.1 - 2 248
AK074767.1 - 2 367
AK074955.1 - 2 362
AK223571.1 - 2 357
AK308873.1 - 2 538
AY339380.1 - 2 376
AY341344.1 - 2 562
BC001510.2 - 2 348 3504557
BI833260.1 - 2 535
BQ072611.1 - 2 328

Gene Symbol
GCOM1
Entrez Gene ID
145781
Gene Name
GRINL1A complex locus 1
Gene Aliases
GRINL1A, Gcom2, MYZAP, MYZAP-POLR2M, gcom
Chromosome Location
Chr. 15: 57591904 - 57717557 on Build GRCh38
UniGene ID
Hs.437256
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001285900.3 NP_001272829.1 13 1558
NR_104371.3 15 2348
GenBank mRNA AY353058.1 - 15 2219
JF419331.1 - 13 1448

View Full Product Details