Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_004120.4 | NP_004111.2 | 3, 4 | 640 | |
GenBank mRNA | AK057968.1 | - | 3, 4 | 553 | |
AK226018.1 | - | 3, 4 | 566 | ||
AK314325.1 | - | 3, 4 | 595 | ||
BC022272.1 | - | 3, 4 | 595 | 4688776 | |
BC073163.1 | - | 3, 4 | 577 | 6151056 | |
CR457062.1 | - | 2, 3 | 326 | ||
M55543.1 | - | 3, 4 | 482 |