Gene Transcripts

Gene Symbol
CRTC2
Entrez Gene ID
200186
Gene Name
CREB regulated transcription coactivator 2
Gene Aliases
TORC-2, TORC2
Chromosome Location
Chr. 1: 153947672 - 153958656 on Build GRCh38
UniGene ID
Hs.406392
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_181715.2 NP_859066.1 2 358
XM_005244946.1 XP_005245003.1 2 339
XM_005244949.2 XP_005245006.1 2 216
XM_017000575.1 XP_016856064.1 2 216
XM_017000576.1 XP_016856065.1 2 216
XM_017000577.1 XP_016856066.1 2 345
XM_017000578.1 XP_016856067.1 2 345
GenBank mRNA AK096382.1 - 2 266
AK223559.1 - 2 311
AY360172.1 - 2 329
AY726597.1 - 2 138
BC053562.1 - 2 269 6188068
BI550818.1 - 2 365

View Full Product Details