Gene Transcripts

Gene Symbol
NCR1
Entrez Gene ID
9437
Gene Name
natural cytotoxicity triggering receptor 1
Gene Aliases
CD335, LY94, NK-p46, NKP46
Chromosome Location
Chr. 19: 54906042 - 54938211 on Build GRCh38
UniGene ID
Hs.97084
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001145457.2 NP_001138929.2 3 236
NM_001145458.2 NP_001138930.2 3 236
NM_004829.6 NP_004820.2 3 236
XM_011527528.2 XP_011525830.1 3 344
XM_011527529.2 XP_011525831.1 3 340
XM_011527530.2 XP_011525832.1 3 332
GenBank mRNA AJ001383.1 - 3 237
AJ006121.1 - 3 205
BC064806.1 - 3 223 5218848

View Full Product Details