Gene Transcripts

Gene Symbol
Kiss1
Entrez Gene ID
280287
Gene Name
KiSS-1 metastasis-suppressor
Gene Aliases
kisspeptin, metastatin
Chromosome Location
Chr. 1: 133321887 - 133329722 on Build GRCm38
UniGene ID
Mm.458301
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178260.3 NP_839991.2 2 460
GenBank mRNA AB162440.1 - 2 422
AB666166.1 - 3 620
AF472576.1 - 2 472

View Full Product Details