Gene Transcripts

Gene Symbol
Speg
Entrez Gene ID
11790
Gene Name
SPEG complex locus
Gene Aliases
AW125581, Apeg1, BPEG, D1Bwg1450e, SPEGalpha, SPEGbeta, mKIAA1297
Chromosome Location
Chr. 1: 75375297 - 75432320 on Build GRCm38
UniGene ID
Mm.275397
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001085370.1 NP_001078839.1 15 3495
NM_007463.4 NP_031489.4 17 4395
GenBank mRNA AF215896.1 - 17 4395
AK147475.1 - 15 3500

View Full Product Details