Gene Transcripts

Gene Symbol
Ccr3
Entrez Gene ID
12771
Gene Name
chemokine (C-C motif) receptor 3
Gene Aliases
CC-CKR3, CKR3, Cmkbr1l2, Cmkbr3
Chromosome Location
Chr. 9: 124013629 - 124031689 on Build GRCm38
UniGene ID
Mm.57050
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009914.4 NP_034044.3 2 2448
GenBank mRNA AK089895.1 - 2 2320
AK153518.1 - 2 2450

View Full Product Details