Gene Transcripts
- Gene Symbol
- Ppargc1a
- Entrez Gene ID
- 19017
- Gene Name
- peroxisome proliferative activated receptor, gamma, coactivator 1 alpha
- Gene Aliases
- A830037N07Rik, ENSMUSG00000079510, Gm11133, PGC-1, PGC-1v, Pgc-1alpha, Pgc1, Pgco1, Ppargc1
- Chromosome Location
- Chr. 5: 51454249 - 52115958 on Build GRCm38
- UniGene ID
- Mm.259072
- Species
- Mus musculus
- Gene Symbol
- Asic2
- Entrez Gene ID
- 11418
- Gene Name
- acid-sensing (proton-gated) ion channel 2
- Gene Aliases
- ACIC2, Accn1, BNC1, BNaC1, BNaC1a, Mdeg
- Chromosome Location
- Chr. 11: 80880163 - 81968396 on Build GRCm38
- UniGene ID
- Mm.234998
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
EF410098.1 |
- |
|
520 |
|
View Full Product Details