Gene Transcripts

Gene Symbol
Xirp1
Entrez Gene ID
22437
Gene Name
xin actin-binding repeat containing 1
Gene Aliases
AI415219, Cmya1
Chromosome Location
Chr. 9: 120013755 - 120023598 on Build GRCm38
UniGene ID
Mm.10117
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011724.3 NP_035854.3 2 954
GenBank mRNA AF051945.2 - 2 925
BC117959.1 - 2 859 40098186
BC117960.1 - 2 859 40098187

View Full Product Details