Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_026456.4 | NP_080732.1 | 4 | 275 | |
GenBank mRNA | AK011757.1 | - | 4 | 283 | |
AK011958.1 | - | 4 | 295 | ||
AK012909.1 | - | 4 | 276 | ||
AK013089.1 | - | 4 | 292 | ||
AK039746.1 | - | 4 | 276 | ||
BC009104.1 | - | 4 | 275 | 3483215 | |
BC028545.1 | - | 4 | 257 | 1399237 |