Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001198570.1 | NP_001185499.1 | 2 | 353 | |
NM_001198571.1 | NP_001185500.1 | 2 | 353 | ||
NM_198127.2 | NP_937760.1 | 2 | 353 | ||
GenBank mRNA | AK081803.1 | - | 2 | 192 | |
BC056345.1 | - | 2 | 174 | 5705048 | |
BC079545.1 | - | 2 | 238 | 6851145 | |
BC079646.1 | - | 2 | 277 | 6850720 |