Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_144913.3 | NP_659162.3 | 4 | 3137 | |
GenBank mRNA | AK008403.1 | - | 1 | 533 | |
AK139731.1 | - | 4 | 1369 | ||
AK154415.1 | - | 4 | 3100 | ||
BC017157.1 | - | 4 | 1406 | ||
BC026876.1 | - | 4 | 2606 | 3662118 | |
BF323293.1 | - | 1 | 248 |