Gene Transcripts

Gene Symbol
Arhgap35
Entrez Gene ID
232906
Gene Name
Rho GTPase activating protein 35
Gene Aliases
6430596G11Rik, AI841135, Grlf1, mKIAA1722, p190A, p190RhoGAP
Chromosome Location
Chr. 7: 16494473 - 16614993 on Build GRCm38
UniGene ID
Mm.28646
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172739.4 NP_766327.3 7 8030
GenBank mRNA AK032559.1 - 1 2783
AK136766.1 - 6 4708
AK137250.1 - 78

View Full Product Details