Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_030229.4 | NP_084505.2 | 4, 5 | 673 | |
GenBank mRNA | AK019868.2 | - | 4, 5 | 674 | |
AK032967.1 | - | 1307 | |||
AK152517.1 | - | 4, 5 | 646 | ||
BC010793.1 | - | 4, 5 | 676 | 4221321 |