Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_133997.2 | NP_598758.1 | 2 | 1360 | |
GenBank mRNA | BC010815.1 | - | 2 | 622 | |
BC022795.1 | - | 2 | 659 | 5102555 | |
BC024769.1 | - | 2 | 647 | 5123598 | |
BC025827.1 | - | 2 | 634 | 5101712 |