Gene Transcripts

Gene Symbol
Abca7
Entrez Gene ID
27403
Gene Name
ATP-binding cassette, sub-family A (ABC1), member 7
Gene Aliases
ABCX, Abc51
Chromosome Location
Chr. 10: 79996453 - 80015572 on Build GRCm38
UniGene ID
Mm.103351
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013850.1 NP_038878.1 1, 2 72
GenBank mRNA AF287141.1 - 1, 2 72
BC145120.1 - 1, 2 111 9053668
BC150708.1 - 1, 2 111 9087619

View Full Product Details