Gene Transcripts

Gene Symbol
CCR6
Entrez Gene ID
1235
Gene Name
C-C motif chemokine receptor 6
Gene Aliases
BN-1, C-C CKR-6, CC-CKR-6, CCR-6, CD196, CKR-L3, CKRL3, CMKBR6, DCR2, DRY6, GPR29, GPRCY4, STRL22
Chromosome Location
Chr. 6: 167111807 - 167139141 on Build GRCh38
UniGene ID
Hs.46468
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_004367.5 NP_004358.2 3 3033
NM_031409.3 NP_113597.2 3 3296

View Full Product Details