Gene Transcripts

Gene Symbol
CNTNAP2
Entrez Gene ID
26047
Gene Name
contactin associated protein-like 2
Gene Aliases
AUTS15, CASPR2, CDFE, NRXN4, PTHSL1
Chromosome Location
Chr. 7: 146116035 - 148420998 on Build GRCh38
UniGene ID
Hs.655684
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014141.5 NP_054860.1 24 7084
GenBank mRNA AF319045.1 - 24 6706
AK000960.1 - 1 2000
CR933671.1 - 4 3263

View Full Product Details