Gene Transcripts
- Gene Symbol
- KIR3DL1
- Entrez Gene ID
- 3811
- Gene Name
- killer cell immunoglobulin like receptor, three Ig domains and long cytoplasmic tail 1
- Gene Aliases
- CD158E1, KIR, KIR3DL1/S1, KIR3DL2, NKAT-3, NKAT3, NKB1, NKB1B
- Chromosome Location
- Chr. 19: 54816438 - 54830778 on Build GRCh38
- UniGene ID
- Hs.645228
- Species
- Homo sapiens
- Gene Symbol
- 1060P11.3
- Entrez Gene ID
- 100506173
- Gene Name
- killer cell immunoglobulin-like receptor, three domains, pseudogene
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Hs.690615
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AJ630586.1 |
- |
5 |
721 |
|
- Gene Symbol
- KIR2DL2
- Entrez Gene ID
- 3803
- Gene Name
- killer cell immunoglobulin like receptor, two Ig domains and long cytoplasmic tail 2
- Gene Aliases
- CD158B1, CD158b, NKAT-6, NKAT6, p58.2
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Hs.743949
- Species
- Homo sapiens
- Gene Symbol
- KIR2DL3
- Entrez Gene ID
- 3804
- Gene Name
- killer cell immunoglobulin like receptor, two Ig domains and long cytoplasmic tail 3
- Gene Aliases
- CD158B2, CD158b, GL183, KIR-023GB, KIR-K7b, KIR-K7c, KIR2DS3, KIR2DS5, KIRCL23, NKAT, NKAT2, NKAT2A, NKAT2B, p58
- Chromosome Location
- Chr. 19: 54738509 - 54753052 on Build GRCh38
- UniGene ID
- Hs.654608
- Species
- Homo sapiens
- Gene Symbol
- KIR2DS2
- Entrez Gene ID
- 100132285
- Gene Name
- killer cell immunoglobulin like receptor, two Ig domains and short cytoplasmic tail 2
- Gene Aliases
- 183ActI, CD158J, CD158b, KIR-2DS2, NKAT-5, NKAT5, cl-49
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Hs.512572
- Species
- Homo sapiens
- Gene Symbol
- KIR2DS3
- Entrez Gene ID
- 3808
- Gene Name
- killer cell immunoglobulin like receptor, two Ig domains and short cytoplasmic tail 3
- Gene Aliases
- NKAT7
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Hs.744266
- Species
- Homo sapiens
- Gene Symbol
- KIR2DS5
- Entrez Gene ID
- 3810
- Gene Name
- killer cell immunoglobulin like receptor, two Ig domains and short cytoplasmic tail 5
- Gene Aliases
- CD158G, NKAT9
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Hs.744266
- Species
- Homo sapiens
- Gene Symbol
- KIR3DS1
- Entrez Gene ID
- 3813
- Gene Name
- killer cell immunoglobulin like receptor, three Ig domains and short cytoplasmic tail 1
- Gene Aliases
- CD158E2, KIR-123FM, KIR-G1, NKAT-10, NKAT10
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Hs.743496
- Species
- Homo sapiens
View Full Product Details