Gene Transcripts
- Gene Symbol
- BIRC5
- Entrez Gene ID
- 332
- Gene Name
- baculoviral IAP repeat containing 5
- Gene Aliases
- API4, EPR-1
- Chromosome Location
- Chr. 17: 78214196 - 78225635 on Build GRCh38
- UniGene ID
- Hs.744872
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001012271.1 |
NP_001012271.1
|
4, 5 |
521 |
|
NM_001168.2 |
NP_001159.2
|
3, 4 |
452 |
|
GenBank mRNA |
AB028869.1 |
- |
4, 5 |
426 |
|
AF077350.1 |
- |
3, 4 |
355 |
|
AK223428.1 |
- |
3, 4 |
397 |
|
AK311917.1 |
- |
3, 4 |
356 |
|
BC008718.2 |
- |
3, 4 |
364 |
2961114 |
BC034148.1 |
- |
3, 4 |
375 |
4656567 |
BC065497.1 |
- |
3, 4 |
345 |
5394399 |
BE537971.1 |
- |
4, 5 |
432 |
|
BI520186.1 |
- |
4, 5 |
531 |
|
BQ687404.1 |
- |
3, 4 |
229 |
|
CB112371.1 |
- |
4, 5 |
454 |
|
CR541740.1 |
- |
3, 4 |
331 |
|
HM625836.1 |
- |
3, 4 |
371 |
|
View Full Product Details