Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_002172.2 | NP_002163.2 | 1 | 616 | |
GenBank mRNA | BC074956.2 | - | 1 | 616 | |
BC104159.1 | - | 1 | 614 | 40029547 | |
BC104160.1 | - | 1 | 614 | 40029549 | |
V00542.1 | - | 1 | 628 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_021057.2 | NP_066401.2 | 1 | 612 | |
GenBank mRNA | BC074991.2 | - | 1 | 610 | |
BC074992.2 | - | 1 | 610 | 30915307 | |
BC113994.1 | - | 1 | 612 | 40084821 | |
BC114000.1 | - | 1 | 612 | 40084820 | |
M34913.1 | - | 1 | 612 |