Gene Transcripts

Gene Symbol
CCL3L3
Entrez Gene ID
414062
Gene Name
C-C motif chemokine ligand 3 like 3
Gene Aliases
464.2, D17S1718, G0S19-2, LD78, LD78BETA, SCYA3L, SCYA3L1
Chromosome Location
Chr. 17: 36194869 - 36196748 on Build GRCh38
UniGene ID
Hs.454713
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001001437.3 NP_001001437.2 2 218
GenBank mRNA AI302038.1 - 2 119
BC007783.2 - 2 213 4308472
BC146919.1 - 2 179 9056847
BC147008.1 - 2 179 9056936
BC147013.1 - 2 179 9056941
BT006783.1 - 2 143
CF128666.1 - 2 197

Gene Symbol
CCL3
Entrez Gene ID
6348
Gene Name
C-C motif chemokine ligand 3
Gene Aliases
G0S19-1, LD78ALPHA, MIP-1-alpha, MIP1A, SCYA3
Chromosome Location
Chr. 17: 36088256 - 36090160 on Build GRCh38
UniGene ID
Hs.514107
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_002983.2 NP_002974.1 2 242
GenBank mRNA AF043339.1 - 1 74
BC071834.1 - 2 214 5935287
BC171831.1 - 2 176 9054485
BC171891.1 - 2 176 9054545
BX103455.1 - 2 242
D00044.1 - 2 223
M23452.1 - 2 223
M25315.1 - 2 223
X03754.1 - 2 223

Gene Symbol
CCL3L1
Entrez Gene ID
6349
Gene Name
C-C motif chemokine ligand 3 like 1
Gene Aliases
464.2, D17S1718, G0S19-2, LD78, LD78-beta(1-70), LD78BETA, MIP1AP, SCYA3L, SCYA3L1
Chromosome Location
Chr. : - on Build
UniGene ID
Hs.512683
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021006.5 NP_066286.1 2 228
NR_111964.1 2 420
GenBank mRNA AV739733.1 - 2 272
BC027888.1 - 2 216 5222247
BC107710.1 - 2 232 6699133
BC146914.1 - 2 179 9056842
X52149.1 - 2 406

View Full Product Details