Gene Transcripts

Gene Symbol
NR3C1
Entrez Gene ID
2908
Gene Name
nuclear receptor subfamily 3 group C member 1
Gene Aliases
GCCR, GCR, GCRST, GR, GRL
Chromosome Location
Chr. 5: 143277931 - 143435512 on Build GRCh38
UniGene ID
Hs.122926
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000176.2 NP_000167.1 9 2731
NM_001018074.1 NP_001018084.1 9 2561
NM_001018075.1 NP_001018085.1 9 2464
NM_001018076.1 NP_001018086.1 9 2357
NM_001018077.1 NP_001018087.1 9 3233
NM_001024094.1 NP_001019265.1 9 2734
NM_001204258.1 NP_001191187.1 9 2731
NM_001204259.1 NP_001191188.1 9 2731
NM_001204260.1 NP_001191189.1 9 2731
NM_001204261.1 NP_001191190.1 9 2731
NM_001204262.1 NP_001191191.1 9 2731
NM_001204263.1 NP_001191192.1 9 2731
NM_001204264.1 NP_001191193.1 9 2731
XM_005268419.3 XP_005268476.1 9 2464
XM_005268420.4 XP_005268477.1 9 2645
XM_005268422.3 XP_005268479.1 9 2526
XM_005268423.3 XP_005268480.1 9 2515
XM_011537637.2 XP_011535939.1 8 3166
XM_017009397.1 XP_016864886.1 9 2357
XM_017009398.1 XP_016864887.1 9 2656
GenBank mRNA AB307716.1 - 8 2239
AK223594.1 - 9 2585
AK302050.1 - 8 1150
AK314789.1 - 9 2361
AM183262.1 - 9 2174
BC015610.2 - 9 2679 4810424
BX640610.1 - 9 2422
BX648553.1 - 10 2526
HQ450643.1 - 8 2239
HQ692822.1 - 8 2239
JN797823.1 - 7 2764
JN797824.1 - 7 2765
U01351.1 - 9 2735
X03225.1 - 9 2371

View Full Product Details