Gene Transcripts

Gene Symbol
ARHGEF28
Entrez Gene ID
64283
Gene Name
Rho guanine nucleotide exchange factor 28
Gene Aliases
RGNEF, RIP2, p190RHOGEF
Chromosome Location
Chr. 5: 73626158 - 73941993 on Build GRCh38
UniGene ID
Hs.482521
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001080479.2 NP_001073948.2 26, 27 3566
NM_001177693.1 NP_001171164.1 26, 27 3566
NM_001244364.1 NP_001231293.1 18, 19 2522
XM_005248568.4 XP_005248625.1 18, 19 4003
XM_011543546.1 XP_011541848.1 26, 27 3573
XM_011543547.2 XP_011541849.1 25, 26 3800
XM_017009702.1 XP_016865191.1 25, 26 3251
XM_017009703.1 XP_016865192.1 17, 18 2304
GenBank mRNA AK025470.1 - 2, 3 282
AK025816.1 - 3, 4 462
AK304761.1 - 18, 19 2522
BC012946.1 - 3, 4 440
BC157846.1 - 25, 26 3412 9057065
BC171850.1 - 25, 26 3412 9054504
CR936816.1 - 4, 5 569

View Full Product Details