Gene Transcripts
- Gene Symbol
- HNRNPK
- Entrez Gene ID
- 3190
- Gene Name
- heterogeneous nuclear ribonucleoprotein K
- Gene Aliases
- AUKS, CSBP, HNRPK, TUNP
- Chromosome Location
- Chr. 9: 83968083 - 83980782 on Build GRCh38
- UniGene ID
- Hs.522257
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001318186.1 |
NP_001305115.1
|
3, 4 |
278 |
|
NM_001318187.1 |
NP_001305116.1
|
3, 4 |
278 |
|
NM_001318188.1 |
NP_001305117.1
|
3, 4 |
278 |
|
NM_002140.4 |
NP_002131.2
|
3, 4 |
439 |
|
NM_031262.3 |
NP_112552.1
|
3, 4 |
439 |
|
NM_031263.3 |
NP_112553.1
|
3, 4 |
278 |
|
XM_005251960.2 |
XP_005252017.1
|
3, 4 |
257 |
|
XM_005251963.3 |
XP_005252020.1
|
3, 4 |
291 |
|
XM_005251965.2 |
XP_005252022.1
|
3, 4 |
249 |
|
XM_011518616.1 |
XP_011516918.1
|
3, 4 |
291 |
|
XM_017014668.1 |
XP_016870157.1
|
3, 4 |
257 |
|
XM_017014669.1 |
XP_016870158.1
|
3, 4 |
249 |
|
GenBank mRNA |
AB209562.1 |
- |
3, 4 |
207 |
|
AB451263.1 |
- |
1, 2 |
52 |
|
AB451390.1 |
- |
1, 2 |
52 |
|
AK096385.1 |
- |
3, 4 |
215 |
|
AK291336.1 |
- |
3, 4 |
261 |
|
AK294067.1 |
- |
3, 4 |
251 |
|
AK300744.1 |
- |
3, 4 |
221 |
|
AY911506.1 |
- |
2, 3 |
115 |
|
BC000355.2 |
- |
2, 3 |
174 |
2964383 |
BC014980.1 |
- |
3, 4 |
245 |
4906241 |
BC025321.1 |
- |
3, 4 |
197 |
|
CR456771.1 |
- |
1, 2 |
52 |
|
DA394419.1 |
- |
3, 4 |
276 |
|
DC318847.1 |
- |
3, 4 |
278 |
|
DC407458.1 |
- |
3, 4 |
311 |
|
DC428989.1 |
- |
3, 4 |
439 |
|
S74678.1 |
- |
2, 3 |
159 |
|
X72727.1 |
- |
3, 4 |
261 |
|
View Full Product Details