Gene Transcripts

Gene Symbol
ARHGAP27
Entrez Gene ID
201176
Gene Name
Rho GTPase activating protein 27
Gene Aliases
CAMGAP1, PP905, SH3D20, SH3P20
Chromosome Location
Chr. 17: 45393902 - 45432916 on Build GRCh38
UniGene ID
Hs.205326
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_174919.3 NP_777579.2 4 1340
GenBank mRNA AF258593.1 - 5 2282
AK290650.1 - 4 1291
BC067345.1 - 4 1340 6084901

View Full Product Details